CAS RN 28049-61-8
CAS RN 28049-61-8
IUPAC Name:
1-(4-chlorophenyl)cyclobutane-1-carbonitrile
Synonyms:
1-(4-chlorophenyl)cyclobutane-1-carbonitrile
Molecular Weight (g/mol):
191.66
Molecular Formula:
C11H10ClN
InChi Key:
XQONXPWVIZZJIL-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1(CCC1)C#N
1
–
4
of
4
results
1
1-(4-Chlorophenyl)cyclobutane Carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(4-Chlorophenyl)cyclobutane Carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(4-Chlorophenyl)cyclobutane Carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1-(4-Chlorophenyl)cyclobutanecarbonitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.