Learn More
1-(4-Chlorophenyl)cyclobutanecarbonitrile, Mikromol™
Product Code. 30049793
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049793 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30049793 Supplier Mikromol Supplier No. MM0991.08
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Chlorophenyl)cyclobutanecarbonitrile
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Chlorophenyl)cyclobutanecarbonitrile
CAS 28049-61-8
Analyte or Component Names 1-(4-Chlorophenyl)cyclobutanecarbonitrile
Shipping Condition Room Temperature
Molecular Formula C11 H10 Cl N
Certifications/Compliance ISO 9001
Synonym 1-(4-Chlorophenyl)cyclobutanecarbonitrile, Sibutramine Hydrochloride Monohydrate Impurity F, Sibutramine Impurity F
InChI Formula InChI=1S/C11H10ClN/c12-10-4-2-9(3-5-10)11(8-13)6-1-7-11/h2-5H,1,6-7H2
SMILES Clc1ccc(cc1)C2(CCC2)C#N
IUPAC Name 1-(4-chlorophenyl)cyclobutane-1-carbonitrile
Molecular Weight (g/mol) 191.66
Quantity 100 mg
Formula Weight 191.0502 g/mol