Learn More
1-(4-Chlorophenyl)cyclobutane Carbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Chlorophenyl)cyclobutanecarbonitrile |
| CAS | 28049-61-8 |
| Molecular Formula | C11 H10 Cl N |
| Quantity | 1 g |
| Synonym | 1-(4-Chlorophenyl)cyclobutanecarbonitrile |
| InChI Formula | InChI=1S/C11H10ClN/c12-10-4-2-9(3-5-10)11(8-13)6-1-7-11/h2-5H,1,6-7H2 |
| SMILES | Clc1ccc(cc1)C2(CCC2)C#N |
| IUPAC Name | 1-(4-chlorophenyl)cyclobutane-1-carbonitrile |
| Molecular Weight (g/mol) | 191.66 |
| Formula Weight | 191.0502 |