CAS RN 2762227-76-7
CAS RN 2762227-76-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl]ethyl]-2-nitroacetamide Sodium Salt, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.