Learn More
N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl]ethyl]-2-nitroacetamide Sodium Salt, Mikromol™
Description
N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl]ethyl]-2-nitroacetamide Sodium Salt
Specifications
Specifications
| Chemical Name or Material | N-[2-[([2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl)ethyl]-2-nitroacetamide Sodium Salt |
| CAS | 2762227-76-7 |
| Analyte or Component Names | N-[2-[([2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulfanyl)ethyl]-2-nitroacetamide Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H17 N4 O3 S2 . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Nizatidine Imp. E (EP), Nizatidine Impurity E as Sodium Salt, Ethanol, 1-[[2-[[[2-[(dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]amino]-2-nitro-, sodium salt (1:1) (ACI) |
| InChI Formula | InChI=1S/C11H18N4O3S2.Na/c1-14(2)6-11-13-9(8-20-11)7-19-4-3-12-10(16)5-15(17)18;/h5,8,12,16H,3-4,6-7H2,1-2H3;/q;+1/p-1 |
| SMILES | [Na+].CN(C)Cc1nc(CSCCNC(=C[N+](=O)[O-])[O-])cs1 |
| IUPAC Name | sodium;1-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethylamino]-2-nitroethenolate |
| Show More |