accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2756-87-8

CH3OOOHO(Z)

CAS RN 2756-87-8

IUPAC Name: (2Z)-4-methoxy-4-oxobut-2-enoic acid
Synonyms: (2Z)-4-methoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol): 130.1
Molecular Formula: C5H6O4
InChi Key: NKHAVTQWNUWKEO-IHWYPQMZSA-N
SMILES: COC(=O)\C=C/C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results