Learn More
Dimethyl Fumarate USP Related Compound A (Monomethyl Fumarate), Mikromol™
Description
Dimethyl Fumarate USP Related Compound A (Monomethyl Fumarate)
Specifications
Specifications
| Chemical Name or Material | Monomethyl Fumarate |
| CAS | 2756-87-8 |
| Analyte or Component Names | Monomethyl Fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H6 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Monomethyl Fumarate, (2E)-2-Butenedioic acid 1-methyl ester, (E)-2-Butenedioic acid monomethyl ester, Fumaric acid methyl ester, Fumaric acid monomethyl ester, Methyl hydrogen fumarate, Monomethyl fumarate |
| InChI Formula | InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+ |
| SMILES | COC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-4-methoxy-4-oxobut-2-enoic acid |
| Show More |