CAS RN 198904-86-8
CAS RN 198904-86-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Des-N-(methoxycarbonyl)-L-tert-leucine Bis-Boc Atazanavir, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Butyl N-[(1S,2S)-1-Benzyl-3-[(tert-butoxycarbonylamino)-[[4-(2-pyridyl)phenyl]methyl]amino]-2-hydroxy-propyl]carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Des-N-(methoxycarbonyl)-L-tert-leucine Bis-Boc Atazanavir, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.