Learn More
Des-N-(methoxycarbonyl)-L-tert-leucine Bis-Boc Atazanavir, TRC
Specifications
Specifications
| Chemical Name or Material | Des-N-(methoxycarbonyl)-L-tert-leucine Bis-Boc Atazanavir |
| CAS | 198904-86-8 |
| Molecular Formula | C32 H42 N4 O5 |
| Quantity | 50 mg |
| Synonym | tert-Butyl N-[(1S,2S)-1-benzyl-3-[(tert-butoxycarbonylamino)-[[4-(2-pyridyl)phenyl]methyl]amino]-2-hydroxy-propyl]carbamate, 2-[(2S,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-4-phenylbutyl]-2-[[4-(2-pyridinyl)phenyl]methyl]-hydrazinecarboxylic acid-1,1-dimethylethyl ester, Hydrazinecarboxylic acid, 2-[3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-4-phenylbutyl]-2-[[4-(2-pyridinyl)phenyl]methyl]-, 1,1-dimethylethyl ester, [S-(R*,R*)]-, 1,1-Dimethylethyl 2-[(2S,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-4-phenylbutyl]-2-[[4-(2-pyridinyl)phenyl]methyl]hydrazinecarboxylate |
| InChI Formula | InChI=1S/C32H42N4O5/c1-31(2,3)40-29(38)34-27(20-23-12-8-7-9-13-23)28(37)22-36(35-30(39)41-32(4,5)6)21-24-15-17-25(18-16-24)26-14-10-11-19-33-26/h7-19,27-28,37H,20-22H2,1-6H3,(H,34,38)(H,35,39)/t27-,28-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)[C@@H](O)CN(Cc2ccc(cc2)c3ccccn3)NC(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-[[(2S,3S)-2-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutyl]-[(4-pyridin-2-ylphenyl)methyl]amino]carbamate |
| Molecular Weight (g/mol) | 562.7 |
| Formula Weight | 562.3155 |