CAS RN 1809427-20-0
CAS RN 1809427-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S,2S,3R)-DT-061, MedChemExpress
MedChemExpress (1S,2S,3R)-DT-061 is an enantiomer of DT-061. DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis.