Learn More
(1S,2S,3R)-DT-061, MedChemExpress
Product Code. 16465556
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465556 100 mg 100mg
16454738 10 mg 10mg
16417457 5 mg 5mg
16401027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16465556 Supplier Medchem Express Supplier No. HY112929B
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress (1S,2S,3R)-DT-061 is an enantiomer of DT-061. DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis.
(1S,2S,3R)-DT-061 is an enantiomer of DT-061. DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis.
Specifications
Chemical Name or Material (1S,2S,3R)-DT-061
CAS 1809427-20-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H23F3N2O5S
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (240.14 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC=C(OC(F)(F)F)C=C1)(N[C@@H]2[C@@H](O)[C@H](N3C4=C(C=CC=C4)OC5=CC=CC=C35)CCC2)=O
Molecular Weight (g/mol) 520.52
Formula Weight 520.52
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death