CAS RN 15029-30-8
IUPAC Name:
3-oxo-3-(piperidin-1-yl)propanenitrile
Synonyms:
piperidine, 1-(cyanoacetyl)-
Molecular Weight (g/mol):
152.2
Molecular Formula:
C8H12N2O
InChi Key:
ANLQHFYDQPMDJY-UHFFFAOYSA-N
SMILES:
O=C(CC#N)N1CCCCC1
1
–
1
of
1
results
1
3-Oxo-3-(piperidin-1-yl)propanenitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.