Learn More
3-Oxo-3-(piperidin-1-yl)propanenitrile, Mikromol™
Description
3-Oxo-3-(piperidin-1-yl)propanenitrile
Specifications
Specifications
| Chemical Name or Material | 3-Oxo-3-(piperidin-1-yl)propanenitrile |
| CAS | 15029-30-8 |
| Analyte or Component Names | 3-Oxo-3-(piperidin-1-yl)propanenitrile |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H12 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Piperidinepropanenitrile, β-oxo-, 1-Piperidinepropionitrile, β-oxo- (5CI), Piperidine, 1-(cyanoacetyl)- (6CI,8CI,9CI), β-Oxo-1-piperidinepropanenitrile, 1-(Cyanoacetyl)piperidine, 3-Oxo-3-(piperidin-1-yl)propanenitrile, Cyanoacetic acid piperidide, Cyanoacetopiperidide, NSC 102772, Piperidinocarbonylacetonitrile |
| InChI Formula | InChI=1S/C8H12N2O/c9-5-4-8(11)10-6-2-1-3-7-10/h1-4,6-7H2 |
| SMILES | O=C(CC#N)N1CCCCC1 |
| IUPAC Name | 3-oxo-3-(1-piperidyl)propanenitrile |
| Show More |