accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 140-40-9

CH3ONHSNNOO

CAS RN 140-40-9

IUPAC Name: N-(5-nitro-1,3-thiazol-2-yl)acetamide
Synonyms: 2-acetamido-5-nitrothiazole
Molecular Weight (g/mol): 187.17
Molecular Formula: C5H5N3O3S
InChi Key: UJRRDDHEMZLWFI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC=C(S1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results