Learn More
Nithiamide, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Nithiamide is a non-5-nitroimidazole drugs, is a antibiotic used in veterinary.
Specifications
Specifications
| Chemical Name or Material | Nithiamide |
| Physical Form | Solid |
| CAS | 140-40-9 |
| Color | Khaki |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H5N3O3S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | CL-5279 Aminitrozole |
| Solubility Information | DMSO : 100 mg/mL (534.25 mM; Need ultrasonic) |
| Show More |