Learn More
VPRBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | VPRBP |
|---|---|
| Concentration | 253 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16878803
|
Proteintech
11612-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16868803
|
Proteintech
11612-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The VPRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPRBP. This antibody recognizes human, mouse, rat antigen. The VPRBP antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
| VPRBP | |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 253 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 9730 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.