Learn More
VPRBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11612-1-AP-20UL
 View more versions of this product
Product Code. 16878803
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The VPRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPRBP. This antibody recognizes human, mouse, rat antigen. The VPRBP antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
VPRBP
Polyclonal
Unconjugated
20 μL
9730
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry
253 μg/mL
Rabbit
Primary
Human, Rat, Mouse