Learn More
VPRBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11612-1-AP-150UL
Description
The VPRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPRBP. This antibody recognizes human, mouse, rat antigen. The VPRBP antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
| VPRBP | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 9730 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 253 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.