missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur-Free Phosphorus Concentrate: P @ approx. 11 wt% in Hydrocarbons, VHG™
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Sulfur-Free Phosphorus Concentrate: P @ approx. 11 wt% in Hydrocarbons
Specifications
Specifications
| Chemical Name or Material | Phosphorus |
| CAS | 7723-14-0 |
| Methods | XRF |
| Analyte or Component Names | Phosphorus |
| Molecular Formula | P |
| Synonym | P |
| InChI Formula | InChI=1S/P |
| SMILES | [P] |
| IUPAC Name | phosphorus |
| Solvent or Matrix | Hydrocarbon Oil |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction