Learn More
Sodium Standard in Base Oil 75, 5000μg/g, SPEX CertiPrep™
Product Code. 11480299 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480299 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11480299 Supplier SPEX Certiprep Supplier No. ORGNA84Z
 Restricted Item
In Stock
Add to basket
Add to basket
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Na5000μg/g
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging Triple-leached LDPE bottle
CAS 7440-23-5, 8042-47-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula Na
MDL Number MFCD00085307
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Na]
IUPAC Name sodium
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 22.99
PubChem CID 5360545
Quantity 50 g
ChEBI CHEBI:26708
Chemical Name or Material Sodium Oil Standard
Show More Show Less
Product intended for laboratory use only