missing translation for 'onlineSavingsMsg'
Learn More

Sodium (Na) Standard Solution, 1000 μg/g in Hydrocarbon Oil, VHG

Product Code. 30328731
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328731 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328731 Supplier VHG Labs Supplier No. VHGONA100050G
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium standard, hydrocarbon oil matrix, elemental calibration standard, ICP/AAS calibration

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-23-5
Analyte or Component Names Sodium
Shipping Condition Room Temperature
Concentration 1000 μg/g
Certifications/Compliance ISO 17034
Synonym Natrium, Sodio, Sodium metal;
InChI Formula InChI=1S/Na
Solution Type Single Solution
SMILES [Na]
IUPAC Name Sodium
Solvent or Matrix 75 cSt Mineral Oil
Industry Type Petrochemical Testing
Quantity 50 g
Concentration or Composition (by Analyte or Components) Sodium 1000 μg/g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.