missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaminase kidney isoform, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaminase kidney isoform, mitochondrial, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_001243239.1 |
---|---|
Gene ID (Entrez) | 2744 |
Name | Glutaminase kidney isoform, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16070770
|
enQuireBio™
QP6095-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090770
|
enQuireBio™
QP6095-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16080770
|
enQuireBio™
QP6095-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000780
|
enQuireBio™
QP6095-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010780
|
enQuireBio™
QP6095-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020780
|
enQuireBio™
QP6095-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001243239.1 | |
Glutaminase kidney isoform, mitochondrial Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
2744 | |
Research Use Only | |
GLS | |
Recombinant Protein | |
E. coli | |
YVGFSNATFQ SERESGDRNF AIGYYLKEKK CFPEGTDMVG ILDFYFQLCS IEVTCESASV MAATLANGGF CPITGERVLS PEAVRNTLSL MHSCGMYDFS GQFAFHVGLP AKSGVAGGIL LVVPNVMGMM CWSPPLDKMG NSVKGIHFCH DLVSLCNFHN YDNLRHFAKK LDPRREGGDQ RVKSVINLLF AAYTGDVSAL RRFALSAMDM EQRDYDSRTA LHVAAAEGHV EVVKFLLEAC KVNPFPKDRW NNTPMDEALH FGHHDVFKIL QEYQVQYTPQ GDSDNGKENQ TVHKNLDGLL | |
Greater than 90% as determined by SDS-PAGE. |