missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaminase kidney isoform, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaminase kidney isoform, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6095-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243239.1 | |
Glutaminase kidney isoform, mitochondrial Protein | |
Research Use Only | |
GLS | |
Recombinant Protein | |
E. coli | |
YVGFSNATFQ SERESGDRNF AIGYYLKEKK CFPEGTDMVG ILDFYFQLCS IEVTCESASV MAATLANGGF CPITGERVLS PEAVRNTLSL MHSCGMYDFS GQFAFHVGLP AKSGVAGGIL LVVPNVMGMM CWSPPLDKMG NSVKGIHFCH DLVSLCNFHN YDNLRHFAKK LDPRREGGDQ RVKSVINLLF AAYTGDVSAL RRFALSAMDM EQRDYDSRTA LHVAAAEGHV EVVKFLLEAC KVNPFPKDRW NNTPMDEALH FGHHDVFKIL QEYQVQYTPQ GDSDNGKENQ TVHKNLDGLL | |
Greater than 90% as determined by SDS-PAGE. |
2744 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |