Learn More
(R)-Amino-(4-hydroxyphenyl)acetic Acid Methyl Ester Hydrochloride, TRC
Product Code. 17927364
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17927364 5 g 5g
17938344 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17927364 Supplier Toronto Research Chemicals Supplier No. TRCA6115555G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57591-61-4
Molecular Formula C9 H11 N O3 . Cl H
Molecular Weight (g/mol) 217.65
Synonym (αR)-α-Amino-4-hydroxy-benzeneacetic Acid Methyl Ester Hydrochloride, α-Amino-(4-hydroxyphenyl)acetic Acid Methyl Ester Hydrochloride, α-Amino-4-hydroxy-benzeneacetic Acid Methyl Ester Hydrochloride, (R)-α-Amino-4-hydroxy-benzeneacetic Acid Methyl Ester Hydrochloride, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride (1:1), (αR)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride, (R)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride, (αR)- (9CI), (R)-Amino(4-hydroxyphenyl)acetic acid methyl ester hydrochloride, (R)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride, D-4-Hydroxyphenylglycine methyl ester hydrochloride, Methyl D-2-p-hydroxyphenylglycine-hydrochloride, Methyl D-4-hydroxyphenylglycine hydrochloride, D-p-Hydroxyphenylglycine Methyl Ester Hydrochloride, HPGMe HCl
IUPAC Name methyl (2R)-2-amino-2-(4-hydroxyphenyl)acetate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)c1ccc(O)cc1
Specifications
Analyte or Component Names (R)-Amino-(4-hydroxyphenyl)acetic Acid Methyl Ester Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C9H11NO3.ClH/c1-13-9(12)8(10)6-2-4-7(11)5-3-6;/h2-5,8,11H,10H2,1H3;1H/t8-;/m1./s1
Solution Type Neat
Industry Type Forensics
Quantity 5 g
Formula Weight 217.0506
Chemical Name or Material (R)-Amino-(4-hydroxyphenyl)acetic Acid Methyl Ester Hydrochloride