accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 57591-61-4

H3COOOHNH2HCl

CAS RN 57591-61-4

IUPAC Name: methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride
Synonyms: methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride
Molecular Weight (g/mol): 217.65
Molecular Formula: C9H12ClNO3
InChi Key: UYCKVJUNDXPDJH-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results