missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBLD Monoclonal Antibody (OTI2B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
POA
Specifications
| Antigen | PBLD |
|---|---|
| Clone | OTI2B5 |
| Concentration | 0.38 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17525271
|
Origene Technologies
TA501824S |
30 μL |
POA
30µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17505271
|
Origene Technologies
TA501824 |
100 μL |
POA
100µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PBLD Monoclonal antibody specifically detects PBLD in Canine, Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PBLD | |
| 0.38 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Primary |
| OTI2B5 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title