Learn More
PBLD Monoclonal Antibody (OTI2B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501824S
Description
PBLD Monoclonal antibody specifically detects PBLD in Canine, Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PBLD | |
| Monoclonal | |
| 0.38 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96PZ0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100856353, 54517 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2B5 | |
| Unconjugated | |
| PUS7 | |
| 0610038K03Rik; epididymis secretory protein Li 306; HEL-S-306; Mawbp; Mawbp2; MAWD binding protein; MAWD binding protein homolog 1; MAWD-binding protein; MAWDBP; Pbld; Pbld1; phenazine biosynthesis like protein domain containing; phenazine biosynthesis-like domain-containing protein; Phenazine biosynthesis-like domain-containing protein 1; phenazine biosynthesis-like protein domain containing; phenazine biosynthesis-like protein domain containing 1; similar to MAWD binding protein (Unknown protein 32 from 2D-page of liver tissue); sphenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue | |
| Full length human recombinant protein of human PUS7 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.