Learn More
PBLD Monoclonal Antibody (OTI2B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501824
Description
The function of the PBLD protein remains unknown.Specifications
| PBLD | |
| Monoclonal | |
| 0.38 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30039 | |
| PBLD | |
| Full length human recombit protein of human PBLD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2B5 | |
| Unconjugated | |
| PBLD | |
| HEL-S-306, MAWBP, MAWDBP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64081 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.