missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otoferlin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
209.00 € - 534.00 €
Specifications
| Antigen | Otoferlin |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18029843
|
Novus Biologicals
NBP1-46574-0.025ML |
0.025 mL |
209.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205975
|
Novus Biologicals
NBP1-46574 |
0.1 mL |
534.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Otoferlin Polyclonal specifically detects Otoferlin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Otoferlin | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| 9381 | |
| IgG | |
| Unpurified | |
| Specific for Otoferlin. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| AUNB1, DFNB6, DFNB9, EC 2.7.10.1, EC 4.2.1.11, FER1L2NSRD9, Fer-1-like protein 2, otoferlin | |
| OTOF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title