missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otoferlin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46574-0.025ML
This item is not returnable.
View return policy
Description
Otoferlin Polyclonal specifically detects Otoferlin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Otoferlin | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| OTOF | |
| A synthetic peptide from n-terminal cytoplasmic region of human Otoferlin (FER1L2) conjugated to an immunogenic carrier protein was used as the antigen. | |
| 0.025 mL | |
| Primary | |
| Specific for Otoferlin. | |
| Rat, Mouse, Human | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1: 300 - 1: 2000, Immunohistochemistry-Paraffin 1 : 300- 1 : 2000 | |
| AUNB1, DFNB6, DFNB9, EC 2.7.10.1, EC 4.2.1.11, FER1L2NSRD9, Fer-1-like protein 2, otoferlin | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 9381 | |
| Reconstitute with 0.1 ml sterilized water to desired concentration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction