missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otoferlin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46574
This item is not returnable.
View return policy
Description
Otoferlin Polyclonal specifically detects Otoferlin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Otoferlin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| OTOF | |
| A synthetic peptide from amino acid region 350-400 of human Otoferlin conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and mouse. | |
| 0.1 mL | |
| Primary | |
| Specific for Otoferlin. | |
| Human, Mouse, Rat | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000 | |
| AUNB1, DFNB6, DFNB9, EC 2.7.10.1, EC 4.2.1.11, FER1L2NSRD9, Fer-1-like protein 2, otoferlin | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 9381 | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction