missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ Meliseptol™ HBV Tissues
Fast-acting surface disinfection wipes for resistant surfaces.
7.00 € - 53.45 €
Specifications
pH | ∼7 |
---|---|
Composition | 180 g Meliseptol solution |
Disposable | Yes |
For Use With (Application) | Rapid disinfection |
Quantity | 100 Wipes |
Description
- Practical and quick with high quality wipes
- For wipe disinfection of small alcohol-resistant surfaces
- Impregnated with Meliseptol rapid liquid
- Broad efficacy spectrum – bactericidal (including MRSA), tuberculocidal and fungicidal (including molds)
- Dispenser box for practical, safe and hygienic removal
- Reusable box with refilling pack
- Dermatologically tested
- Save-lock with audible click
- 90 days of in-use stability
- Active against enveloped viruses (including HBV, HIV, HCV) and non-enveloped viruses (including Norovirus)
- Exposure time of only 1 minute for enveloped viruses, 10 minutes for non-enveloped viruses
- Fresh smell
Specifications
∼7 | |
Yes | |
100 Wipes |
180 g Meliseptol solution | |
Rapid disinfection |