missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Meliseptol™ HBV Tissues

Product Code. 15427924
Change view
Click to view available options
Container Type:
Dispenser box
Refilling pack
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type unitSize
15427924 Dispenser box Each
15447924 Refilling pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427924 Supplier B Braun™ Supplier No. 18706

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fast-acting surface disinfection wipes for resistant surfaces.

  • Practical and quick with high quality wipes
  • For wipe disinfection of small alcohol-resistant surfaces
  • Impregnated with Meliseptol rapid liquid
  • Broad efficacy spectrum – bactericidal (including MRSA), tuberculocidal and fungicidal (including molds)
  • Dispenser box for practical, safe and hygienic removal
  • Reusable box with refilling pack
  • Dermatologically tested
  • Save-lock with audible click
  • 90 days of in-use stability
  • Active against enveloped viruses (including HBV, HIV, HCV) and non-enveloped viruses (including Norovirus)
  • Exposure time of only 1 minute for enveloped viruses, 10 minutes for non-enveloped viruses
  • Fresh smell

Specifications

pH ∼7
Composition 180 g Meliseptol solution
Container Type Dispenser box
Disposable Single-use
For Use With (Application) Rapid disinfection
Quantity 100 Wipes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.