missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Meliseptol™ HBV Tissues

Product Code. 15447924
Change view
Click to view available options
Container Type:
Dispenser box
Refilling pack
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Unit Size
15447924 Refilling pack Pack of 100
15427924 Dispenser box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447924 Supplier B Braun™ Supplier No. 18707
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fast-acting surface disinfection wipes for resistant surfaces.

  • Practical and quick with high quality wipes
  • For wipe disinfection of small alcohol-resistant surfaces
  • Impregnated with Meliseptol rapid liquid
  • Broad efficacy spectrum – bactericidal (including MRSA), tuberculocidal and fungicidal (including molds)
  • Dispenser box for practical, safe and hygienic removal
  • Reusable box with refilling pack
  • Dermatologically tested
  • Save-lock with audible click
  • 90 days of in-use stability
  • Active against enveloped viruses (including HBV, HIV, HCV) and non-enveloped viruses (including Norovirus)
  • Exposure time of only 1 minute for enveloped viruses, 10 minutes for non-enveloped viruses
  • Fresh smell

Specifications

pH ∼7
Composition 180 g Meliseptol solution
Container Type Refilling pack
Disposable Single-use
For Use With (Application) Rapid disinfection
Quantity 100 Wipes
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Meliseptol HBV Tissues
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Propane-1-ol, Didecyldimethylammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.