Learn More
Lubricating Oil Standards Set (contains one of each LOIL1 thru LOIL16), VHG™
Click to view available options
Quantity:
16 x 100 mL
Unit Size:
1600mL
Description
Lubricating Oil Standards Set (contains one of each LOIL1 thru LOIL16)
Specifications
Specifications
| Chemical Name or Material | Calcium/Phosphorus/Sulfur/Zinc |
| CAS | 7440-70-2, 7723-14-0, 7704-34-9, 7440-66-6 |
| Methods | XRF |
| Analyte or Component Names | Calcium/Phosphorus/Sulfur/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 16 x 100 mL |
| Concentration or Composition (by Analyte or Components) | Calcium 0%/Phosphorus 0%/Sulfur 0% |