missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
4385.00 € - 21200.00 €
Specifications
| Antigen | Tyrosine kinases |
|---|---|
| Packaging | 1 Kit(s) |
| Primary or Secondary | Primary |
| Content And Storage | Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C. |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10617764
|
Invitrogen
PV4844 |
25 μg |
4385.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10717815
|
Invitrogen
PV4845 |
1 mg |
21200.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A part of the JAK/STAT Pathway, STAT1 (Signal Transducer and Activator of Transcription 1) is phosphorylated at Tyr701 by the Janus family of tyrosine kinases.For use in combination with the LanthaScreen™ STAT1 U206 cell line (K1469) and the GFP-STAT1 physiological protein substrate (PV5211) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
| Tyrosine kinases | |
| Primary | |
| RUO |
| 1 Kit(s) | |
| Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C. |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title