missing translation for 'onlineSavingsMsg'
Learn More

LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit

LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit

Brand:  Invitrogen PV4845

 View more versions of this product

Product Code. 10717815

  • 21200.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.

This item is not returnable. View return policy

Description

Description

A part of the JAK/STAT Pathway, STAT1 (Signal Transducer and Activator of Transcription 1) is phosphorylated at Tyr701 by the Janus family of tyrosine kinases.

For use in combination with the LanthaScreen™ STAT1 U206 cell line (K1469) and the GFP-STAT1 physiological protein substrate (PV5211) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tyrosine kinases
0.45mg/mL
150kDa
RUO
Primary
Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C.
384-well Plate
Human
End Point
Tyrosine Kinases
JAK1, JAK2, JAK3, TYK2
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
Tb (Terbium)/GFP (EmGFP)/Lanthanide Chelates
1 mg
Kinase Biology
1 Kit(s)
150000 Da
340⁄490, 520
Dry Ice
Hu
TR-FRET
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.