Learn More
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
Brand:  Invitrogen PV4844
 View more versions of this product
Product Code. 10617764
  • 4385.00 € / 25µg
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
This item is not returnable. View return policy
Description
Description
A part of the JAK/STAT Pathway, STAT1 (Signal Transducer and Activator of Transcription 1) is phosphorylated at Tyr701 by the Janus family of tyrosine kinases.
For use in combination with the LanthaScreen™ STAT1 U206 cell line (K1469) and the GFP-STAT1 physiological protein substrate (PV5211) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tyrosine kinases
Tb (Terbium)/GFP (EmGFP)
25 μg
Protein Assays and Analysis
1 Kit(s)
150000 Da
384-well Plate
Human
End Point
STAT1, JAK1, JAK2 JH1 JH2, JAK2, TYK2, JAK3, JAK2 JH1 JH2 V617F
TR-FRET
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit, 25μg, Tyrosine Kinases Antigen, Human Species Reactivity, Used For Biochemical Kinase Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays,
RUO
Primary
Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C.
Biochemical terbium phospho-specific activity, Cell-based terbium phospho-specific activity (stable)
340⁄490, 520
Dry Ice
Hu
Tyrosine Kinases
JAK1, JAK2, JAK3, TYK2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.