missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF17 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | KIF17 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16843344
|
Proteintech
14615-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16833344
|
Proteintech
14615-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transports vesicles containing N-methyl-D-aspartate (NMDA) receptor 2B along microtubules.Specifications
| KIF17 | |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Non-human Primate, Rat, Mouse, Human | |
| Q99PW8, Q9P2E2 | |
| 16559, 500571, 57576 | |
| KIF17 Fusion Protein Ag6217 | |
| Primary | |
| -20°C | |
| KIF17 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| KIAA1405, KIF17, KIF17B, KIF3 related motor protein, KIF3X, kinesin family member 17, Kinesin like protein KIF17 | |
| Kif17 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title