Learn More
KIF17 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14615-1-AP-20UL
 View more versions of this product
Product Code. 16843344
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Transports vesicles containing N-methyl-D-aspartate (NMDA) receptor 2B along microtubules.
Specifications
Specifications
KIF17
Polyclonal
Unconjugated
KIF17
KIAA1405, KIF17, KIF17B, KIF3 related motor protein, KIF3X, kinesin family member 17, Kinesin like protein KIF17
Rabbit
Antigen Affinity Chromatography
RUO
16559, 500571, 57576
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99PW8, Q9P2E2
Kif17
KIF17 Fusion Protein Ag6217
20 μL
Primary
Non-human Primate, Rat, Mouse, Human