missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF17 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14615-1-AP-150UL
This item is not returnable.
View return policy
Description
Transports vesicles containing N-methyl-D-aspartate (NMDA) receptor 2B along microtubules.Specifications
| KIF17 | |
| Polyclonal | |
| Unconjugated | |
| KIF17 | |
| KIAA1405, KIF17, KIF17B, KIF3 related motor protein, KIF3X, kinesin family member 17, Kinesin like protein KIF17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16559, 500571, 57576 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99PW8, Q9P2E2 | |
| Kif17 | |
| KIF17 Fusion Protein Ag6217 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Non-human Primate, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction