missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MED24 Full-length ORF (NP_055630.2, 1 a.a. - 989 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_055630.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9862 |
Molecular Weight (g/mol) | 136.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184182
|
Abnova™
H00009862-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174182
|
Abnova™
H00009862-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSCHGKAEECIGLCRALLSALHWLLRCTAASAERLREGLEAGTPAAGEKQLAMCLQRLEKTLSSTKNRALLHIAKLEEASSWTAIEHSLLKLGEILANLSNPQLRSQAEQCGTLIRSIPTMLSVHAEQMHKTGFPTVHAVILLEGTMNLTGETQSLVEQLTMVKRMQHIPTPLFVLEIWKACFVGLIESPEGTEELKWTAFTFLKIPQVLVKLKKYSHGDKDFTEDVNCAFEFLLKLTPLLDKADQRCNCDCTNFLLQECGKQGLLSEASVNNLMAKRKADREHAPQQKSGENANIQPNIQLILRAEPTVTNILKTMDADHSKSPEGLLGVLGHMLSGKSLDLLLAAAAATGKLKSFARKFINLNEFTTYGSEESTKPASVRALLFDISFLMLCHVAQTYGSEVILSESRTGAEVPFFETWMQTCMPEEGKILNPDHPCFRPDSTKVESLVALLNNSSEMKLVQMKWHEACLSISAAILEILNAWENGVLAFESIQKITDNIKGKVCSLAVCAVAWLVAHVRMLGLDEREKSLQMIRQLAGPLFSENTLQFYNERVVIMNSILERMCADVLQQTATQIKFPSTGVDTMPYWNLLPPKRPIKEVLTDIFAKVLEKGWVDSRSIHIFDTLLHMGGVYWFCNNLIKELLKETRKEHTLRAVELLYSIFCLDMQQVTLVLLGHILPGLLTDSSKWHSLMDPPGTALAKLAVWCALSSYSSHKGQASTRQKKRHREDIEDYISLFPLDDVQPSKLMRLLSSNEDDANILSSPTDRSMSSSLSASQLHTVNMRDPLNRVLANLFLLISSILGSRTAGPHTQFVQWFMEECVDCLEQGGRGSVLQFMPFTTVSELVKVSAMSSPKVVLAITDLSLPLGRQVAAKAIAALSpecifications
NP_055630.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
136.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSCHGKAEECIGLCRALLSALHWLLRCTAASAERLREGLEAGTPAAGEKQLAMCLQRLEKTLSSTKNRALLHIAKLEEASSWTAIEHSLLKLGEILANLSNPQLRSQAEQCGTLIRSIPTMLSVHAEQMHKTGFPTVHAVILLEGTMNLTGETQSLVEQLTMVKRMQHIPTPLFVLEIWKACFVGLIESPEGTEELKWTAFTFLKIPQVLVKLKKYSHGDKDFTEDVNCAFEFLLKLTPLLDKADQRCNCDCTNFLLQECGKQGLLSEASVNNLMAKRKADREHAPQQKSGENANIQPNIQLILRAEPTVTNILKTMDADHSKSPEGLLGVLGHMLSGKSLDLLLAAAAATGKLKSFARKFINLNEFTTYGSEESTKPASVRALLFDISFLMLCHVAQTYGSEVILSESRTGAEVPFFETWMQTCMPEEGKILNPDHPCFRPDSTKVESLVALLNNSSEMKLVQMKWHEACLSISAAILEILNAWENGVLAFESIQKITDNIKGKVCSLAVCAVAWLVAHVRMLGLDEREKSLQMIRQLAGPLFSENTLQFYNERVVIMNSILERMCADVLQQTATQIKFPSTGVDTMPYWNLLPPKRPIKEVLTDIFAKVLEKGWVDSRSIHIFDTLLHMGGVYWFCNNLIKELLKETRKEHTLRAVELLYSIFCLDMQQVTLVLLGHILPGLLTDSSKWHSLMDPPGTALAKLAVWCALSSYSSHKGQASTRQKKRHREDIEDYISLFPLDDVQPSKLMRLLSSNEDDANILSSPTDRSMSSSLSASQLHTVNMRDPLNRVLANLFLLISSILGSRTAGPHTQFVQWFMEECVDCLEQGGRGSVLQFMPFTTVSELVKVSAMSSPKVVLAITDLSLPLGRQVAAKAIAAL | |
RUO | |
MED24 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9862 | |
MED24 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARC100/CRSP100/CRSP4/DRIP100/KIAA0130/MGC8748/THRAP4/TRAP100 | |
MED24 | |
Recombinant | |
wheat germ expression system |