missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MED24 Full-length ORF (NP_055630.2, 1 a.a. - 989 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009862-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184182

  • 508.00 € / 25µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSCHGKAEECIGLCRALLSALHWLLRCTAASAERLREGLEAGTPAAGEKQLAMCLQRLEKTLSSTKNRALLHIAKLEEASSWTAIEHSLLKLGEILANLSNPQLRSQAEQCGTLIRSIPTMLSVHAEQMHKTGFPTVHAVILLEGTMNLTGETQSLVEQLTMVKRMQHIPTPLFVLEIWKACFVGLIESPEGTEELKWTAFTFLKIPQVLVKLKKYSHGDKDFTEDVNCAFEFLLKLTPLLDKADQRCNCDCTNFLLQECGKQGLLSEASVNNLMAKRKADREHAPQQKSGENANIQPNIQLILRAEPTVTNILKTMDADHSKSPEGLLGVLGHMLSGKSLDLLLAAAAATGKLKSFARKFINLNEFTTYGSEESTKPASVRALLFDISFLMLCHVAQTYGSEVILSESRTGAEVPFFETWMQTCMPEEGKILNPDHPCFRPDSTKVESLVALLNNSSEMKLVQMKWHEACLSISAAILEILNAWENGVLAFESIQKITDNIKGKVCSLAVCAVAWLVAHVRMLGLDEREKSLQMIRQLAGPLFSENTLQFYNERVVIMNSILERMCADVLQQTATQIKFPSTGVDTMPYWNLLPPKRPIKEVLTDIFAKVLEKGWVDSRSIHIFDTLLHMGGVYWFCNNLIKELLKETRKEHTLRAVELLYSIFCLDMQQVTLVLLGHILPGLLTDSSKWHSLMDPPGTALAKLAVWCALSSYSSHKGQASTRQKKRHREDIEDYISLFPLDDVQPSKLMRLLSSNEDDANILSSPTDRSMSSSLSASQLHTVNMRDPLNRVLANLFLLISSILGSRTAGPHTQFVQWFMEECVDCLEQGGRGSVLQFMPFTTVSELVKVSAMSSPKVVLAITDLSLPLGRQVAAKAIAAL
Specifications

Specifications

NP_055630.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
136.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARC100/CRSP100/CRSP4/DRIP100/KIAA0130/MGC8748/THRAP4/TRAP100
MED24
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9862
MED24 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSCHGKAEECIGLCRALLSALHWLLRCTAASAERLREGLEAGTPAAGEKQLAMCLQRLEKTLSSTKNRALLHIAKLEEASSWTAIEHSLLKLGEILANLSNPQLRSQAEQCGTLIRSIPTMLSVHAEQMHKTGFPTVHAVILLEGTMNLTGETQSLVEQLTMVKRMQHIPTPLFVLEIWKACFVGLIESPEGTEELKWTAFTFLKIPQVLVKLKKYSHGDKDFTEDVNCAFEFLLKLTPLLDKADQRCNCDCTNFLLQECGKQGLLSEASVNNLMAKRKADREHAPQQKSGENANIQPNIQLILRAEPTVTNILKTMDADHSKSPEGLLGVLGHMLSGKSLDLLLAAAAATGKLKSFARKFINLNEFTTYGSEESTKPASVRALLFDISFLMLCHVAQTYGSEVILSESRTGAEVPFFETWMQTCMPEEGKILNPDHPCFRPDSTKVESLVALLNNSSEMKLVQMKWHEACLSISAAILEILNAWENGVLAFESIQKITDNIKGKVCSLAVCAVAWLVAHVRMLGLDEREKSLQMIRQLAGPLFSENTLQFYNERVVIMNSILERMCADVLQQTATQIKFPSTGVDTMPYWNLLPPKRPIKEVLTDIFAKVLEKGWVDSRSIHIFDTLLHMGGVYWFCNNLIKELLKETRKEHTLRAVELLYSIFCLDMQQVTLVLLGHILPGLLTDSSKWHSLMDPPGTALAKLAVWCALSSYSSHKGQASTRQKKRHREDIEDYISLFPLDDVQPSKLMRLLSSNEDDANILSSPTDRSMSSSLSASQLHTVNMRDPLNRVLANLFLLISSILGSRTAGPHTQFVQWFMEECVDCLEQGGRGSVLQFMPFTTVSELVKVSAMSSPKVVLAITDLSLPLGRQVAAKAIAAL
RUO
MED24
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions