Learn More
Abnova™ Human MED24 Full-length ORF (NP_055630.2, 1 a.a. - 989 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009862-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSCHGKAEECIGLCRALLSALHWLLRCTAASAERLREGLEAGTPAAGEKQLAMCLQRLEKTLSSTKNRALLHIAKLEEASSWTAIEHSLLKLGEILANLSNPQLRSQAEQCGTLIRSIPTMLSVHAEQMHKTGFPTVHAVILLEGTMNLTGETQSLVEQLTMVKRMQHIPTPLFVLEIWKACFVGLIESPEGTEELKWTAFTFLKIPQVLVKLKKYSHGDKDFTEDVNCAFEFLLKLTPLLDKADQRCNCDCTNFLLQECGKQGLLSEASVNNLMAKRKADREHAPQQKSGENANIQPNIQLILRAEPTVTNILKTMDADHSKSPEGLLGVLGHMLSGKSLDLLLAAAAATGKLKSFARKFINLNEFTTYGSEESTKPASVRALLFDISFLMLCHVAQTYGSEVILSESRTGAEVPFFETWMQTCMPEEGKILNPDHPCFRPDSTKVESLVALLNNSSEMKLVQMKWHEACLSISAAILEILNAWENGVLAFESIQKITDNIKGKVCSLAVCAVAWLVAHVRMLGLDEREKSLQMIRQLAGPLFSENTLQFYNERVVIMNSILERMCADVLQQTATQIKFPSTGVDTMPYWNLLPPKRPIKEVLTDIFAKVLEKGWVDSRSIHIFDTLLHMGGVYWFCNNLIKELLKETRKEHTLRAVELLYSIFCLDMQQVTLVLLGHILPGLLTDSSKWHSLMDPPGTALAKLAVWCALSSYSSHKGQASTRQKKRHREDIEDYISLFPLDDVQPSKLMRLLSSNEDDANILSSPTDRSMSSSLSASQLHTVNMRDPLNRVLANLFLLISSILGSRTAGPHTQFVQWFMEECVDCLEQGGRGSVLQFMPFTTVSELVKVSAMSSPKVVLAITDLSLPLGRQVAAKAIAALSpecifications
NP_055630.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
136.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARC100/CRSP100/CRSP4/DRIP100/KIAA0130/MGC8748/THRAP4/TRAP100 | |
MED24 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9862 | |
MED24 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKVVNLKQAILQAWKERWSDYQWAINMKKFFPKGATWDILNLADALLEQAMIGPSPNPLILSYLKYAISSQMVSYSSVLTAISKFDDFSRDLCVQALLDIMDMFCDRLSCHGKAEECIGLCRALLSALHWLLRCTAASAERLREGLEAGTPAAGEKQLAMCLQRLEKTLSSTKNRALLHIAKLEEASSWTAIEHSLLKLGEILANLSNPQLRSQAEQCGTLIRSIPTMLSVHAEQMHKTGFPTVHAVILLEGTMNLTGETQSLVEQLTMVKRMQHIPTPLFVLEIWKACFVGLIESPEGTEELKWTAFTFLKIPQVLVKLKKYSHGDKDFTEDVNCAFEFLLKLTPLLDKADQRCNCDCTNFLLQECGKQGLLSEASVNNLMAKRKADREHAPQQKSGENANIQPNIQLILRAEPTVTNILKTMDADHSKSPEGLLGVLGHMLSGKSLDLLLAAAAATGKLKSFARKFINLNEFTTYGSEESTKPASVRALLFDISFLMLCHVAQTYGSEVILSESRTGAEVPFFETWMQTCMPEEGKILNPDHPCFRPDSTKVESLVALLNNSSEMKLVQMKWHEACLSISAAILEILNAWENGVLAFESIQKITDNIKGKVCSLAVCAVAWLVAHVRMLGLDEREKSLQMIRQLAGPLFSENTLQFYNERVVIMNSILERMCADVLQQTATQIKFPSTGVDTMPYWNLLPPKRPIKEVLTDIFAKVLEKGWVDSRSIHIFDTLLHMGGVYWFCNNLIKELLKETRKEHTLRAVELLYSIFCLDMQQVTLVLLGHILPGLLTDSSKWHSLMDPPGTALAKLAVWCALSSYSSHKGQASTRQKKRHREDIEDYISLFPLDDVQPSKLMRLLSSNEDDANILSSPTDRSMSSSLSASQLHTVNMRDPLNRVLANLFLLISSILGSRTAGPHTQFVQWFMEECVDCLEQGGRGSVLQFMPFTTVSELVKVSAMSSPKVVLAITDLSLPLGRQVAAKAIAAL | |
RUO | |
MED24 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |