missing translation for 'onlineSavingsMsg'
Learn More

Human IL-22 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 31 publication

289.00 € - 532.00 €

Specifications

Antigen IL-22
Clone 142928
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18741508
GREENER_CHOICE
View Documents
R&D Systems
IC7821P-025
25 Tests
289.00 €
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18703027
GREENER_CHOICE
View Documents
R&D Systems
IC7821P-100
100 Tests
532.00 €
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications

Specifications

IL-22
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
IL22
IgG1
Protein A or G purified from hybridoma culture supernatant
Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10.
142928
Flow Cytometry
R-PE
Mouse
Human
Q9GZX6
50616
E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.