Learn More
Human IL-22 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 31 publication
Brand:  R&D Systems IC7821P-025
 View more versions of this product
Product Code. 18741508
  • 289.00 € / 25 tests
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-22
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
50616
Human
Purified
Flow Cytometry
142928
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Q9GZX6
IL22
E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6
25 Tests
Primary
Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1