missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-22 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 31 publication
Brand: R&D Systems IC7821P-100
This item is not returnable.
View return policy
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-22 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50616 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 142928 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q9GZX6 | |
| IL22 | |
| E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
| 100 Tests | |
| Primary | |
| Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction