Learn More
Hamilton™ Luer Lock Adapters for Hamilton™Valves
Adapters are designed to work with Hamilton valves
18.00 € - 22.00 €
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10271543
|
Hamilton™
35030 |
Adapter w/Male Luer Lock |
22.00 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10313033
|
Hamilton™
35031 |
Adapter w/Female Luer Lock |
18.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
1/4-28 Thread .059 in. (1.5mm) |