Learn More
Hamilton™ Luer Lock Adapters for Hamilton™Valves
Product Code. 10271543
Click to view available options
Type:
Adapter with Female Luer Lock
Adapter with Male Luer Lock
Unit Size:
Each
Product Code. 10271543
Brand: Hamilton™ 35030
Only null left
Add to basket
Add to basket
Adapters are designed to work with Hamilton valves
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
Type Adapter with Male Luer Lock
Thread Size 1/4-28 Thread .059 in. (1.5mm)