Learn More
Hamilton™ Luer Lock Adapters for Hamilton™Valves
Adapters are designed to work with Hamilton valves
Brand: Hamilton™ 35031
Additional Details : Weight : 0.01000kg
Description
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
Adapter w/Female Luer Lock | |
1/4-28 Thread .059 in. (1.5mm) |